missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol (mixture of isomers) 99.0+%, TCI America™
$63.38 - $275.87
Chemical Identifiers
| CAS | 1319-77-3 |
|---|---|
| Molecular Formula | C7H8O |
| MDL Number | MFCD00151099 |
| Synonym | Cresylic Acid, Tricresol, Methylphenol |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C069825ML
|
TCI America
C069825ML |
25 mL |
Each for $63.38
|
|
|||||
|
C0698500ML
|
TCI America
C0698500ML |
500 mL |
Each for $275.87
|
|
|||||
Chemical Identifiers
| 1319-77-3 | |
| C7H8O | |
| MFCD00151099 | |
| Cresylic Acid, Tricresol, Methylphenol |
Specifications
| 1319-77-3 | |
| 192°C | |
| MFCD00151099 | |
| 2022 | |
| 108.14 | |
| Liquid |
| Yellow | |
| C7H8O | |
| 25 mL | |
| Cresylic Acid, Tricresol, Methylphenol | |
| ≥99.0% (GC) | |
| Cresol (mixture of isomers) |
Safety and Handling
EINECSNumber : (3)-0499&(4)-0057
RTECSNumber : GO5950000
TSCA : Yes