missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cresol (mixture of isomers) 99.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 192°C |
| Quantity | 500 mL |
| UN Number | 2022 |
| Formula Weight | 108.14 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Cresol (mixture of isomers) |
Safety and Handling
| EINECSNumber | (3)-0499&(4)-0057 |
| RTECSNumber | GO5950000 |
| TSCA | Yes |