missing translation for 'onlineSavingsMsg'
Learn More

Cresol (mixture of isomers) 99.0+%, TCI America™

Catalog No. C069825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C069825ML 25 mL
C0698500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C069825ML Supplier TCI America Supplier No. C069825ML
Only null left

Chemical Identifiers

CAS 1319-77-3
Molecular Formula C7H8O
MDL Number MFCD00151099
Synonym Cresylic Acid, Tricresol, Methylphenol

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 mL
UN Number 2022
Formula Weight 108.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cresol (mixture of isomers)
EINECSNumber (3)-0499&(4)-0057
RTECSNumber GO5950000
TSCA Yes