missing translation for 'onlineSavingsMsg'
Learn More

Creatinine, 98%, Spectrum™ Chemical

Catalog No. 18609957
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
10 g
100 g
12 kg
25 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18609957 25 g Poly Bottle
18609954 10 g Amber Glass Bottle
18609955 100 g Poly Bottle
18609953 1 kg Poly Pail
18609956 12 kg Poly Pail
18609958 25 kg Fiber Drum
6 options

Catalog No. 18609957

Supplier: Spectrum Chemical Mfg Cor CR11025GM

Only null left

Chemical Identifiers

CAS 60-27-5

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 25 g
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.