missing translation for 'onlineSavingsMsg'
Learn More

Creatine phosphate disodium salt tetrahydrate, 98+%

Catalog No. AAA1536214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1536214 25 g
AAA1536203 1 g
AAA1536206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1536214 Supplier Thermo Scientific Chemicals Supplier No. A1536214
Only null left

CAS: 71519-72-7 | C4H8N3Na2O5P·4H2O

Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.

Solubility
Soluble in water.0.1 g/mL.

Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.

Chemical Identifiers

CAS 71519-72-7
Molecular Formula C4H8N3Na2O5P·4H2O
MDL Number MFCD00150192
Synonym Sodium creatine phosphate dibasic tetrahydrate

Specifications

Assay Percent Range ≥98%
Quantity 25 g
Solubility Information Soluble in water.0.1g/mL.
Formula Weight 327.14 (255.07 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Creatine phosphate disodium salt tetrahydrate
EINECSNumber 213-074-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only