missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine phosphate, disodium salt, 99.12%, MP Biomedicals™
$1101.19 - $1101.19
Chemical Identifiers
| CAS | 922-32-7 |
|---|---|
| Molecular Formula | C4H8N3Na2O5P |
| Molecular Weight (g/mol) | 255.08 |
| MDL Number | MFCD00150192 |
| InChI Key | RNTXMYSPASRLFT-UHFFFAOYSA-L |
| Synonym | creatine phosphate sodium salt |
| PubChem CID | 131887655 |
| IUPAC Name | disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate |
| SMILES | [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10052025
|
MP Biomedicals Inc
0210052025 |
25 g |
Each for $1,101.19
|
|
|||||
Chemical Identifiers
| 922-32-7 | |
| 255.08 | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| 131887655 | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
| C4H8N3Na2O5P | |
| MFCD00150192 | |
| creatine phosphate sodium salt | |
| disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate |
Specifications
| White | |
| Crystalline Powder | |
| 98-100% | |
| MFCD00150192 | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate | |
| 131887655 | |
| Creatine phosphate, disodium salt |
| 922-32-7 | |
| 25 g | |
| C4H8N3Na2O5P | |
| creatine phosphate sodium salt | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O | |
| 255.08 | |
| 327.2 |
Safety and Handling
Recommended Storage : Store at - 0°C