missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate, Hexahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18611438
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611438 50 kg Amber Glass Bottle
18611434 125 g Amber Glass Bottle
18611435 500 g Amber Glass Bottle
18611436 2.5 kg Amber Glass Bottle
18611437 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611438 Supplier Spectrum Chemical Mfg Cor Supplier No. M105050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
IUPAC Name magnesium(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

pH 5.0 to 8.2
Physical Form Crystalline, Liquid
CAS Max % 1
Quantity 50 kg
Packaging Amber Glass Bottle
Percent Purity 98 to 102%
Grade ACS/Reagent
Chemical Name or Material Magnesium Nitrate, Hexahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.