missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Creatine Hydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 295°C |
| Color | White |
| Quantity | 100 g |
| Formula Weight | 131.14 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Creatine Hydrate |
Safety and Handling
| RTECSNumber | MB7706000 |
| TSCA | Yes |