Learn More
Eisco™ Crank and Connecting Rod Mechanical Demonstration Model
The Crank and Connecting Rod is an interactive visual demonstration of a common mechanical linkage system widely used in internal combustion engines.
Supplier:  Eisco™ PH0366TM46
Catalog No. S27673
Only null left
Description
Description
  • Hand-cranked interactive model for visual learning.
  • Shows the fundamentals of how a crank and connecting rod function.
  • Open system is sandwiched by two clear plastic plates for unobstructed study.
  • Constructed with heavy duty metal and plastic.
  • Includes comprehensive instruction and experiment manual.
Specifications
Specifications
Crank and Connecting Rod Mechanical Demonstration Model
Product Suggestions
Product Suggestions