Learn More
Micronova™ CR100 PC™ Cleanroom Tape—Orange
Processed for applications in and around cleanrooms
$1,427.40
Products 1
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Width (English) Price Quantity & Availability  
19035334G
View Documents Promotion Details
Micronova™
CR100PC34OR
75 in.
Case of 48 Each for $1,427.40
Only null left
Description
Description
  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag
Order Info
Available in red, yellow, white, black, brown, dark blue, dark green, light blue, light green, orange, purple, and transparent in 36 yd. long (33m) rolls. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here.