missing translation for 'onlineSavingsMsg'
Learn More

CPC Monohydrate (=Cetylpyridinium Chloride Monohydrate) 98.0+%, TCI America™

Catalog No. a51615g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51615G Supplier TCI America Supplier No. A51615G
Only null left

[Ion association reagent for extraction photometric analysis and sensitizer in photometric determination of metals]

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
IUPAC Name 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

Melting Point 84°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 339.99
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material CPC Monohydrate (=Cetylpyridinium Chloride Monohydrate) [Ion association reagent for extracti
RTECSNumber UU5075000
TSCA Yes