missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ CP-310 Alkaline Detergent

Catalog No. 17010133
Change view
Click to view available options
Size:
5 gal.
55 gal.
Container Type:
Drum
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17010133 5 gal. Pail
17010134 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17010133 Supplier STERIS™ Supplier No. 1K7405

CP-310 Alkaline Cleaner is a high-performance detergent for pharmaceutical, biotech, and cosmetic industries. Formulated with low-mercury potassium hydroxide, surfactants, and chelating agents, it effectively removes process residues. Low foaming, suitable for CIP and COP.

CP-310 Alkaline Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW CP-310 WORKS

CP-310 Alkaline Detergent is formulated with a proprietary blend of low-mercury grade potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

WHY CP-310?

  • Productivity - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Cleaner
Color-coded No
Form Clear Liquid
pH 12.4
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.