Learn More
Neuvitro Corporation Poly-L-lysine coated sterile German glass coverslips for cell culture, 18mm dia #1.5 thick 90/pk, ready to use in cell culture. DFS Item
Supplier:  Neuvitro GG1815PLL
CE certified German glass coverslips with Poly-L-lysine coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with synthetic Poly-L-lysine on both sides and sterile. 18mm diameter round, #1.5 thickness, 90 pieces/pack. Ready to use directly in cell culture. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 0.5 year when stored in chest freezer. User manual included.
Catalog No. NC0326897
Only null left
Explore available promotions