Learn More
Covidien Monoject™ Sterile Syringes
Catalog No. 0556163
Change view
Click to view available options
Volume (Metric):
3 mL
Catalog No. Volume (Metric)
0556163 3 mL
1 options
Catalog No. 0556163 Supplier Covidien Supplier No. 8881513934
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Autoclavable, sterile and latex-free
Specifications
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) For use in O.R./procedure packs
Latex Free Yes
Product Type Monoject™ Standard Syringe
Disposable Single-use
Volume (Metric) 3 mL
Needle Point Style Tri-beveled, Anti-coring
Connection Type Luer Lock
Graduated Yes
Sterility Sterile
Show More Show Less