Learn More
Covidien Monoject™ Silicone-Coated Tubes with Red Stoppers
Standard blood-collection tubes
Supplier:  Covidien 8881301512
 View more versions of this product
Catalog No. 22032264
Only null left
Description
Description
Standard blood-collection tubes
  • Interior coated with water-soluble silicone material
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
  • Sterilized by gamma radiation
  • Used for laboratory procedures requiring serum; general chemistry and serology procedures
Specifications
Chemical Identifiers
Silicone Coated Tube
No Additive
Sterile
Red
Specifications
Silicone Coated Tube
Sterile
Red
No Additive
7 mL
13 x 100 mm
Product Suggestions
Product Suggestions