Learn More
Covidien Monoject™ Rigid Pack 60mL Syringes without Needles
Sterile, single-use, individually packaged syringes ready for use.
Supplier:  Covidien 8881560125
 View more versions of this product
Catalog No. 22-031-375
Only null left
Explore available promotions
Description
Description
  • Autoclavable in rigid pack for use in O.R./procedure packs
  • Latex-free
Specifications
Specifications
Autoclavable
Yes
Single-use
No Needle Included
Luer Lock
For use in O.R./procedure packs
Monoject™ Standard Syringe
0.25 oz.
60 mL
Sterile
Product Suggestions
Product Suggestions