missing translation for 'onlineSavingsMsg'
Learn More

Covidien Monoject™ Nonsilicone-Coated Blood Collection Tubes with Light Blue Stoppers

Catalog No. 22034407
Change view
Click to view available options
Capacity (Metric):
4.5 mL
Closure Color:
Light Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22034407 4.5 mL Light Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22034407 Supplier Covidien Supplier No. 8881340478

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blood collection tubes with sterile interior.

  • Nonsilicone coated tubes
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
  • Sterilized by gamma radiation

Specifications

Product Type Non-silicone Coated Tube
Additive Buffered Sodium Citrate 0.5mL 3.8 pct. (0.129M) Solution
Capacity (Metric) 4.5 mL
Closure Color Light Blue
Tube Size 13 x 75 mm
For Use With (Application) Blood collection