missing translation for 'onlineSavingsMsg'
Learn More

Covidien Monoject™ Noncoated Tubes with Red Stoppers

Catalog No. 22302718
Change view
Click to view available options
Capacity (Metric):
10 mL
Closure Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22302718 10 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22302718 Supplier Covidien Supplier No. 8881302718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No additive, nonsilicone coated tubes used for laboratory procedures

  • Sterile tube interior
  • Stopper coating: glycerine
  • Dimensions: 16 × 100mm (0.63 × 3.94 in.)
  • Capacity: 10mL draw

Laboratory procedures requiring serum; general chemistry and serology procedures

Specifications

Additive No Additive
Capacity (Metric) 10 mL
Closure Color Red
Length (Metric) 100 mm
Diameter (Metric) Outer 16 mm