missing translation for 'onlineSavingsMsg'
Learn More

Covidien Monoject™ Magellan ™ Safety Needle

Catalog No. 22256575
Change view
Click to view available options
Needle Gauge:
23 G
25 G
Length (English) Needle:
1 in.
Color:
Blue
Orange
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Needle Color
22256575 23 G 1 in. Blue
22256577 25 G 1 in. Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22256575 Supplier Covidien Supplier No. 8881850310
Only null left

One-handed operation needle-based safety technology

Specifications

Color Blue
Needle Gauge 23 G
For Use With (Application) Protects healthcare worker from accidental needlestick injuries
Includes Robust safety shield
Latex Free Yes
Length (English) Needle 1 in.
Product Type Magellan™ Hypodermic Safety Needle
Needle Point Style Tri-Bevel
Sterility Sterile