missing translation for 'onlineSavingsMsg'
Learn More

Covidien Maxicare Underpads

Catalog No. 22037950
Change view
Click to view available options
Dimensions (L x W):
24 x 23 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22037950 24 x 23 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22037950 Supplier Covidien Supplier No. FUS7134
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For incontinence and skin care

  • Extra-large size provides maximum coverage, absorption and protection
  • Super-absorbent lining with Poly-Fresh polymer draws fluids away from the surface and neutralizes odors
  • Recommended for use in chairs as well as beds
  • Reduce costs by eliminating the need for multiple underpads

Specifications

Product Type Underpad
Color Light Blue
Backsheet Material Polypropylene
Topsheet Material Spunbond Non-woven
Dimensions (L x W) 24 x 23 in.
For Use With (Application) For incontinence and skin care
For Use With (Equipment) Chairs and Beds