missing translation for 'onlineSavingsMsg'
Learn More

Covidien Defender™ Safety Needle Holders

Catalog No. 22257421
Change view
Click to view available options
Color:
Blue
Purple
Length (English):
1 in.
1.5 in.
Length (Metric):
2.5 cm
3.8 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English) Length (Metric)
22257421 Purple 1.5 in. 3.8 cm
22257420 Blue 1 in. 2.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22257421 Supplier Covidien Supplier No. 8881225115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Total needle protection allows for safe, efficient and easy use

Single-use devices eliminate needle removal and blood tube holder reuse, and help prevent needle-stick injury

  • One-handed safety activation
  • Front and back end needle protection
  • Needle retracts into blood tube holder to reduce sharps disposal space
  • May be used with 1 in. or 1.5 in. standard blood collection needles
  • Latex free

Compliance

Meets FDA Quality System Requirements (QSR) per 21 Code of Federal Regulations (CFR) Part 820. Complies with Occupational Safety and Health Administration (OSHA) Standards.

Specifications

Disposable Single-use
Length (Metric) 3.8 cm
Color Purple
For Use With (Application) Provide front and back end needle protection
Length (English) 1.5 in.
Product Type Needle Holder