missing translation for 'onlineSavingsMsg'
Learn More

Covidien Chemosafety™ Containers

Catalog No. p-3238086
Change view
Click to view available options
Capacity (English):
2 gal.
8 gal.
18 gal.
Closure Type:
Hinged Top
Slide Top
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Closure Type
14830137 8 gal. Hinged Top
14830136 2 gal. Hinged Top
14830117 18 gal. Slide Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14830137 Supplier Covidien Supplier No. 8985
Only null left

For disposal of chemotherapy waste

  • Yellow color makes container highly identifiable as chemotherapy waste
  • Accommodates assorted sizes of waste materials and sharps, including syringes, ampules, and prep materials
  • Sealing gasketed lid provides leak-resistant containment of fluid
  • Container locks to make it secure for final disposal
  • Molded handles aid in secure transport
  • Nestable containers save valuable storage space

Specifications

Capacity (Metric) 30.2 L
Closure Type Hinged Top
Capacity (English) 8 gal.
Color Yellow
For Use With (Application) For disposal of trace chemotherapy waste
Material Polypropylene
Product Type Sharps Disposal Container