Learn More
Neuvitro CE certified German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1 thick 0.5oz/pk untreated
Supplier:  Neuvitro GG12OZ
Encompass
CE certified cell culture tested German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1 thickness 0.5oz/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.
Numéro de catalogue. NC0512913
Il en reste null