Learn More
Electron Microscopy German Glass Coverslips #1, 18 mm DFS Item
Supplier:  Electron Microscopy Sciences 7229007
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 18 mm
Catalog No. 50-192-9521
Only null left
Explore available promotions