missing translation for 'onlineSavingsMsg'
Learn More

StatLab™ CoverSafe Mounting Medium

Catalog No. STLMMC0243
Change view
Click to view available options
Volume (Metric):
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
STLMMC0243 500 mL
STLMMC0226 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLMMC0243 Supplier StatLab™ Supplier No. MMC0243

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or inhaled.

A fast-drying, low-hazard, optically clear, permanent mounting medium.

  • CoverSafe contains no xylene, no toluene, no dioxane, and no benzene
  • Pleasant citrus scent, making it an ideal low-hazard permanent mounting medium
  • For use in histology and cytology

Specifications

Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 500 mL