missing translation for 'onlineSavingsMsg'
Learn More

Uniform Technology Coverall Ultimax Grid, White

Cleanroom coveralls feature a military collar, raglan sleeves, knit cuffs, and a coil zipper with top snaps.

$104.55 - $104.55

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Polyester
Color White
Disposable Reusable
View More Specs

Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
01-670-748
View Documents
Uniform Technology
CC1245TD09WHXSS
X-Small
Each for $104.55
Only null left
 
Description

Description

  • Ideal for General CE Spaces/Applications (Non-ESD sensitive Medical Device, Electronics & Aerospace, Food Processing/Handling or Packaging, Cultivation Industry, University Labs, Imaging/Film coating Manufactures, General Medical/Dental Facilities)
  • Coverall
  • Ultimax Grid
  • Military Collar
  • Raglan Sleeves
  • Knit Cuffs
  • Coil Zipper with Top Snaps
Specifications

Specifications

Cleanroom Coveralls
Polyester
Reusable
Knit
Zipper
Non-sterile
Military
1
1/Ea.
Unisex
White
Snap
Collared
Bound
Anti-static
Yes
Raglan
Class 10 (ISO 4)
Videos
SDS
Documents

Documents