missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enespro™ Enespro™ ArcGuard™ 40 cal Coverall
Protective clothing that provides the highest level of protection in work with electrical exposures and hazards
Supplier: Enespro™ C88UQUQ3X32
Description
- Recoil action allows full range of motion
- All metal covered to ensure no skin contact
- Chest pockets with flaps and concealed snap closures
- Comfortable elastic sewn waist
- Convenient side pockets for access to trouser pockets
- FR fabric will NOT melt, drip or ignite
- Two front pockets with step to access trouser pockets
Specifications
| Coveralls | |
| Unisex | |
| Navy | |
| Elastic (Stitched-in) | |
| Collared | |
| FR Hook and Loop | |
| Non-sterile | |
| Flame | |
| NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269 | |
| Coveralls |
| 3X-Large | |
| Cotton, Nylon | |
| Reusable | |
| Open | |
| Unhooded | |
| Stitched | |
| CAT 4 | |
| 40 cal/cm² | |
| None |