Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 12 cal Protera Coverall
Coveralls with two-way covered zipper down front for easy entry
$557.63 - $557.63
Specifications
Product Type Coveralls
Gender Unisex
Material DuPont™ Protera™
Color Navy
Disposable Reusable
View More Specs
Products 1
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17001416
View Documents
National Safety Apparel™
C88LYXL32
X-Large
Each for $557.63
Only null left
Description
Description
  • Constructed from Protera™
  • Comfortable, sewn-in elastic waist
  • Two-way covered zipper down front for easy entry
  • Breast pockets with flaps and hidden snap closures
  • Two large rear pockets
  • Two front pockets with pass-through slashes for access to pants pockets
  • All metal covered to ensure no contact
Specifications
Specifications
Coveralls
DuPont™ Protera™
Reusable
Open
Unhooded
Stitched
CAT 2
12 cal/cm²
None
Unisex
Navy
Elastic (Stitched-in)
Collared
Zipper
Non-sterile
Flame
ASTM F1506, NFPA 70E, CSA Z462, OSHA 1910.269
Coveralls
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'