Learn More
Electron Microscopy Circular Cover Glass #1.5, 12 mm, 597 Pieces/oz 1 OZ DFS Item
Supplier:  Electron Microscopy Sciences 7223001
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 597 Pieces/oz
Thickness: 0.16-0.19 mm
Catalog No. 50949008
Only null left
Explore available promotions