Learn More
Electron Microscopy Circular Cover Glass #1.5, 12 mm, 597 Pieces/oz 10 oz
Supplier: Electron Microscopy Sciences 7223010
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.Diameter: 12 mmPackage: 597 Pieces/ozThickness: 0.16-0.19 mm