Learn More
Therapak™ Courier Shipping Systems
For variable temperature shipments by ground or air
Supplier:  Therapak™ 36520
 View more versions of this product
Catalog No. 22130211
Only null left
Description
Description
  • For sample shipments that require temperature integrity from 24 to 96 hours—ideal for international shipments
  • Reusable urethane shippers with durable, thick wall construction provide more than twice the R-value of EPS shippers
  • Courier tote can be configured to accommodate various configurations, including: three small shippers, two medium shippers, one large shipper, or two small and one medium shipper
  • Replacement lids available separately
Specifications
Chemical Identifiers
For specimen transport
Urethane
Specifications
3 small, 2 medium, 1 large; or 2 small and 1 medium shippers
3 Sm, 2 Med, 1 Lg or 2 Sm and 1 Med
Urethane
For specimen transport
Courier Tote
Product Suggestions
Product Suggestions