Learn More
Therapak™ Courier Shipping Systems
For variable temperature shipments by ground or air
Supplier: Therapak™ 36517
Description
- For sample shipments that require temperature integrity from 24 to 96 hours—ideal for international shipments
- Reusable urethane shippers with durable, thick wall construction provide more than twice the R-value of EPS shippers
- Courier tote can be configured to accommodate various configurations, including: three small shippers, two medium shippers, one large shipper, or two small and one medium shipper
- Replacement lids available separately
Chemical Identifiers
| For specimen transport | |
| Urethane |
Specifications
| For specimen transport | |
| Small | |
| Shipper | |
| Urethane |