Learn More
Corning™ Counting Chamber
Catalog No. 07200988
Change view
Click to view available options
Grid Type:
Gridless
Catalog No. Grid Type
07200988 Gridless
1 options
Catalog No. 07200988 Supplier Corning™ Supplier No. 480200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Corning counting chamber is a reusable glass cell counting slide for the Corning cell counter.
This gridless slide has a 0.1 mm depth same as a Neubauer Hemocytometer and comes standard with two high quality cover slips. These slides have a notch for easy loading of cell suspension.
Specifications
Grid Type Gridless
Depth (Metric) 0.1 mm
For Use With (Equipment) Corning™ Cell Counter