missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Countess™ Cell Counting Chamber Slides

Catalog No. C10283
Change view
Click to view available options
Type:
Cell Counting Chamber Slide
Reusable Slide
Includes:
No Stain
SafeCount Stain, 2 x 1 mL Vials
Trypan Blue Stain, 100 mL Bottle
Trypan Blue Stain, 2 x 1 mL Vials
Trypan Blue Stain, 2 x 100 mL Bottles
Trypan Blue Stain, 20 x 1 mL Vials
Quantity:
1 Glass Slide
1250 Slides
2500 Slides
50 Slides
500 Slides
5000 Slides
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Includes Quantity
C10283 Cell Counting Chamber Slide No Stain 50 Slides
A25750 Reusable Slide No Stain 1 Glass Slide
A40008469 Cell Counting Chamber Slide SafeCount Stain, 2 x 1 mL Vials 50 Slides
C10228 Cell Counting Chamber Slide Trypan Blue Stain, 2 x 1 mL Vials 50 Slides
C10312 Cell Counting Chamber Slide Trypan Blue Stain, 20 x 1 mL Vials 500 Slides
C10313 Cell Counting Chamber Slide Trypan Blue Stain, 100 mL Bottle 1250 Slides
C10314 Cell Counting Chamber Slide Trypan Blue Stain, 100 mL Bottle 2500 Slides
C10315 Cell Counting Chamber Slide Trypan Blue Stain, 2 x 100 mL Bottles 5000 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. C10283 Supplier Invitrogen™ Supplier No. C10283

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Countess Cell Counting Chamber Slides are conveniently packaged for use with the Countess Automated Cell Counters. Each disposable and reusable slide features two enclosed chambers.

The Countess Chamber Slides are designed for use with the Countess 3 and 3FL automated cell counters.

The disposable Countess Cell Counting Chamber Slides offer convenience in automated cell counting for processing multiple cell samples, busy cell culture facilities, and easy disposal of contaminated or hazardous samples. They also eliminate the risk of contamination from residual samples. After use, Countess slides can be disposed of as standard biohazard waste.

The Countess Reusable Slide is a green and cost-efficient alternative to disposable slides for automated cell counting. The reusable slide enables researchers and cell culture facilities to reduce costs and generate less waste per year, making it affordable for labs to automate their cell counts. The reusable slide is cleaned and dried between counts, just as with a hemocytometer.

Each Countess Cell Counting Chamber Slide has two separate, enclosed chambers for counting one sample in duplicate or two different samples. Cell counting occurs in a central location of the counting chamber. The volume counted is 0.4 μL, the same as counting four 1-mm2 squares in a standard hemocytometer.

The Countess Automated Cell Counter works in three simple steps:

  • Mix 10 μL of sample with 10 μL of a cell viability stain or other desired reagent, including ReadyCount™ stains, and pipet into a Countess chamber slide
  • Insert the slide into the instrument
  • Press 'Count'

That's it—get the total cell count, percent viable cells, and average cell size in as little as 30 seconds.

Specifications

Type Cell Counting Chamber Slide
Product Line Countess
For Use With (Equipment) Countess Automated Cell Counter, Trypan Blue Stain (Cat. No. T10282)
Includes No Stain
Content And Storage • Countess™ Cell Counting Slides, 1 x 50 slides

Note: Trypan blue stain is not included
Quantity 50 Slides
Do you offer an alternative for Countess Cell Counting Chamber Slides (Cat. no. C10283) for Countess 3 and Countess 3 FL Automated Cell Counter?

Yes. The alternative options for Countess Cell Counting Chamber Slides (Cat. No. C10283) and for Countess 3 and Countess 3 FL Automated Cell Counter can be found using the link below:

Countess Cell Counting Chamber Slides (Cat. Nos. C10228, C10312, C10313, C10314, C10315)

For Research Use Only. Not for use in diagnostic procedures.