missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Countess™ 3 FL Automated Cell Counter

Catalog No. p-200076305
Encompass
Change view
Click to view available options
Type:
Automated Cell Counter
Automated Cell Counter Starter Package
Warranty:
1-year Standard
3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage)
3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement)
Software:
Countess 3/3 FL Cell Counter Software
Countess 3/3 FL Cell Counter Software with 21 CFR Part 11 Software Module
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Warranty Software
A49866 Automated Cell Counter Starter Package 1-year Standard Countess 3/3 FL Cell Counter Software
A49892 Automated Cell Counter 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement) Countess 3/3 FL Cell Counter Software
A49893 Automated Cell Counter 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage) Countess 3/3 FL Cell Counter Software
A50299 Automated Cell Counter Starter Package 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement) Countess 3/3 FL Cell Counter Software
A50303 Automated Cell Counter Starter Package 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage) Countess 3/3 FL Cell Counter Software
AMQAF2000 Automated Cell Counter 1-year Standard Countess 3/3 FL Cell Counter Software
AMQAF2001 Automated Cell Counter 1-year Standard Countess 3/3 FL Cell Counter Software with 21 CFR Part 11 Software Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. A49866 Supplier Invitrogen™ Supplier No. A49866

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience rapid cell counting with the Countess 3 FL Automated Cell Counter. This brightfield and fluorescence cell counter has advanced optics, automated focus and lighting, and AI-based image analysis software for counting cells and nuclei, monitoring fluorescence, and measuring cell viability.

The Countess 3 FL Automated Cell Counter is a benchtop brightfield and fluorescence cell counter with advanced optics, automated focus and lighting, and image analysis software driven by a neural network-based machine-learning algorithm for rapid automated cell counting of primary and immortalized cell lines. The Countess 3 FL counter allows brightfield or fluorescence cell counting with three-channel flexibility (brightfield and two optional fluorescence channels), enabling researchers to count cells and isolated nuclei, monitor fluorescence, examine apoptosis, and measure cell viability.

Features of the Countess 3 FL Automated Cell Counter include:

  • Accurate—highly accurate machine-learning algorithm applied to automated cell counting, eliminates the subjectivity of manual cell counting and user-to-user variability
  • Flexible—accepts up to two EVOS light cubes, user selectable from fifteen fluorescent channels; compatible with both disposable and reusable cell counting chamber slides
  • Fast—counts live and dead cells, measures viability, reports average cell size, calculates percent cell aggregates, and saves data with a single touch, in about 20 seconds
  • Convenient—requires no cleaning, routine maintenance, or calibration; high-resolution capacitive touchscreen and simple user interface provide quick startup and requires minimal training
  • Versatile—perform automated counting of single cells and isolated nuclei for RNA sequencing and gene expression applications in single-cell analysis workflows

The Countess 3 FL Automated Cell Counter works in two simple steps:

  1. Mix 10 μL of sample with 10 μL of trypan blue or other desired reagent, including ReadyCount stain, and pipet into a disposable or reusable Countess chamber slide.
  2. Insert the slide into the instrument. Rapid Capture and Auto-Save modes allow automated cell counting with a single touch. Focus, lighting, and file saving are executed automatically.

Obtain accurate cell counts

The Countess 3 FL Automated Cell Counter uses trypan blue staining combined with auto-focus and lighting driven by a machine-learning image analysis algorithm to obtain accurate cell and viability counts. The measurement range extends from 1 x 104 to 1 x 107 cells/mL (the optimal range is 1 x 105 to 4 x 106 cells/mL), which is broader than that of a hemocytometer.

Simple operation with fluorescent application versatility

Operated using an intuitive user interface, automated cell counting with the Countess 3 FL counter gives you the data you need to prepare your sample for the next passage or experiment and allows you to multiplex with a variety of fluorescent dyes and proteins excited and detected via EVOS light cubes. With both brightfield and fluorescence cell counting options, you can perform cell counts for routine cell culture applications along with more extensive fluorescence analysis of cells such as live vs. dead, nucleated vs. non-nucleated, and transfected vs. untransfected. You no longer need complex, expensive instruments to obtain reliable data.

Transfer data and images to your PC or cloud

Using the USB port on the Countess 3 FL Automated Cell Counter and any USB drive, you can save your cell count results and images and transfer them to your PC. For your convenience, a USB drive and WiFi dongle are supplied with the Countess 3 FL instrument.

Improve downstream single-cell analysis

Ensure accurate single-cell analysis and RNA sequencing results by minimizing cell aggregates and obtaining precise viable cell counts. For optimal single-cell data output, use the percent cell aggregates on the Countess 3 FL to confirm <5% aggregates in the starting sample before single-cell analysis. Staining with fluorescent dyes such as the ReadyCount stains and using the Countess 3 FL-based count mode offers more accurate counts and viability measurements for samples with debris or out-of-focus cells in brightfield. The Countess 3 FL and ReadyCount stains are also recommended for counting isolated nuclei in scATAC-Seq studies. View the single cell and nuclei counting application note for more details.

The Countess 3 FL Automated Cell Counter can be purchased as the instrument (Cat. No. AMQAF2000) or a variety of packages bundled with additional accessories, extended warranties, or software, such as the Countess 3 FL Starter Package (Cat. No. A49866), which includes:

  • Countess FL Automated Cell Counter
  • EVOS GFP and TexasRed light cubes
  • Reusable slide
  • 500 disposable slides with trypan blue

Extended warranty instrument and starter packages

The Countess 3 FL Automated Cell Counter comes with a standard 1-year warranty that covers all costs for shipping, parts, and labor for repairs. Service plans can be bundled with new instrument purchases in an extended warranty package that extends the standard warranty by 2 years for a total of 3. Additional years of coverage can also be purchased to extend the standard 1-year or extended 3-year warranty. The Countess 3 FL instrument and starter packages are offered with two different types of bundled extended warranty packages:

  • 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement)
    • Replacement shipped out in 1 business day for labs with critical timelines
    • Available with the instrument only (Cat. No. A49892) and the Countess 3 FL Starter Package (Cat. No. A50299)
  • 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage)
    • Instrument repaired and returned in ~3 weeks for regulated labs required to maintain asset tagging consistency (loaner instrument shipped within 1 business day with upgrade to an ABRC Support Plus contract)
    • Available with the instrument only (Cat. No. A49893) and the Countess 3 FL Starter Package (Cat. No. A50303)

Countess 3 FL with 21 CFR Part 11 Software Module (Cat. No. AMQAF2001)

The included Countess 3 21 CFR Part 11 Software Module is for use with the Countess 3 and Countess 3 FL automated cell counters and supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using an SAE administration console. The 21 CFR Part 11 software can also be purchased separately (Cat. No. A51025).

Specifications

Type Automated Cell Counter Starter Package
Product Line Countess
Camera 5 MP
For Use With (Application) Cell Culture and Analysis Workflows
For Use With (Equipment) EVOS Microscopes, Countess Chamber Slides, and Reusable Slide
Detection Method Brightfield, Fluorescence
Holds 2 samples per slide
Magnification 2.5X
Optics 3 Channels (Brightfield and 2 slots for EVOS™ LED light cubes)
Display LCD (1280 x 800 pixels)
Memory Onboard, USB drive
Software Countess 3/3 FL Cell Counter Software
Dimensions 9 x 9 x 5.5 in. (228.6 x 228.6 x 139.7 mm)
Weight (English) 8 lb.
Weight (Metric) 3.6 kg
Certifications/Compliance RoHS
Content And Storage
  • Countess 3 FL Automated Cell Counter, 1 each
  • Power cord with 4 adapters (for US/Canada/Taiwan/Japan, Europe, or UK), 1 each
  • Countess Cell Counting Chamber Slides, 50 slides/box
  • Countess 3 FL Disposable Slide Holder, 1 each
  • Countess 3 FL Reusable Slide Holder, 1 each
  • Countess 3 FL Light Cube Removal Tool, 1 each
  • Countess 3 USB Drive, 1 each
  • Countess 3 FL Automated Cell Counter Quick Reference Card, 1 each
  • Wi-Fi Dongle (Cat. No. A26774), 1 each
  • EVOS Light Cube, GFP 2.0 (Cat. No. AMEP4951), 1 each
  • EVOS Light Cube, TexasRed 2.0 (Cat. No. AMEP4955), 1 each
  • Countess Reusable Slide (Cat. No. A25750), 1 each
  • Disposable Countess Cell Counting Chamber Slides (Cat. No. C10283), 10 x 50 slides
  • Trypan blue (Cat. No. T10282), 20 x 1 mL vials

Store at room temperature.

Quantity 1 package
Shipping Condition Ambient
Warranty 1-year Standard
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
Where can I find the service retirement letter for the Countess II and II FL Automated Cell Counters?

You can find the service retirement letter for Countess II and II FL Automated Cell Counters below:

Important end-of-life communication for Countess II and II FL products

Where can I find FAQs for Countess II and II FL Automated Cell Counters?

Countess II and II FL Automated Cell Counter FAQs can be found in our Product FAQ Database.

Where can I find a troubleshooting information for Countess 3 and 3 FL Automated Cell Counters?

Troubleshooting information for Countess 3 and 3FL can be found on the Cell Counting Support Center.

What is the shelf life for Countess Cell Counting Chamber Slides, disposable?

Our guarantee period for disposable Countess Cell Counting Chamber Slides is 5 years.

How can I find information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter?

For information on the regulatory and compliance standards for my Countess 3/3FL Automated Cell Counter, including 21 CFR Part 11, IQ/OQ, and warranty packages, please see refer to Countess Automated Cell Counter Services on our website.

Where can I find safety and handling information for my Countess 3/3FL Automated Cell Counter?

For safety and handling information for your Countess 3/3FL Automated Cell Counter, please refer to “Appendix E Safety” in the User Guide.

Countess 3 Automated Cell Counter User Guide (Pub. No. MAN0019566 E.0)

How can I interpret the data and graphs generated by my Countess 3/3FL Automated Cell Counter?

Please refer to the following link for many helpful Countess Automated Cell Counter demo videos, found under the Resources tab, covering such topics as "How to Visualize histograms and perform gating.” Our Cell Analysis Resource center contains additional educational resources:

Countess Automated Cell Counter Resources

What are the step-by-step instructions to correctly set up my Countess 3/3FL Automated Cell Counter out of the box?

For user manuals, instructional videos, and educational content for Countess Automated Cell Counter instruments, please consult the following link:

Countess Automated Cell Counter Resources

How can I request a certificate of analysis for my Countess 3/3FL Automated Cell Counter?

Please search our Documents and Certificates database for your COA, using the specific serial number for your instrument. If your search is unsuccessful, please go to Contact Us, click on Technical Support, enter your product name, and complete the form to request your COA, or contact technical support at Technicalservices@thermofisher.com.

How does the accuracy of my Countess 3/3FL Automated Cell Counter compare with manual cell counting methods?

Compared to manual hemocytometer counts with Countess 3/3FL Automated Cell Counter instruments can reduce data errors and variability from many causes, including operator subjectivity and fatigue, counting area, and challenging samples. Countess 3 cell counters achieve both the accuracy of manual cell counts and the precision of flow cytometer counts, without the expense or training required by flow cytometry.

Find more information about the accuracy and precision of cell counting with Countess 3 instruments using the links below:

Countess Automated Cell Counter Features

Cell counting accuracy and precision: Why it matters and how to achieve it (White Paper)

What improvements and new features does my Countess 3/3FL Automated Cell Counter offer compared to earlier versions of the Countess?

Relative to the Countess II, the Countess 3/3FL Automated Cell Counter offers the following core improvements:
•     Increased accuracy and precision
•     Artificial intelligence deep-learning algorithm
•     Rapid capture & autosave
•     Fewer touch points; time-savings

How can I troubleshoot inaccurate cell counting results on my Countess 3/3FL Automated Cell Counter?

Please consult the following link for basic troubleshooting tips: Cell Counting Information for more information. If the above fails to correct the problem, please contact technical support at Technicalservices@thermofisher.com.

How can I use my Countess 3/3FL Automated Cell Counter with different cell types, such as mammalian cells and bacterial cells?

Countess instruments are compatible with a broad range of cell types and have been verified for use with several commonly used cell lines and primary cell types. Bacteria (and particles under 5 microns in diameter) may require fluorescent mode for counting and may not be detected in brightfield mode.

How should I interpret the cell counting results displayed by Countess 3/3FL Automated Cell Counter?

The Countess offers several ways to save count data. The Report PDF is a convenient summary of count results that includes cell concentration, fluorescence detected (for the properly equipped Countess 3FL only), and images of the counting field. Users may also export data in CSV, FCS, and image formats.

How can I optimize the cell counting process on my Countess 3/3FL Automated Cell Counter for accuracy and speed?

Users may create customized protocols for individual workflows or sample types. Please refer to Chapter 3 "Protocols" in the User Guide for more information.

Countess 3 Automated Cell Counter User Guide (Pub. No. MAN0019566 E.0)

How should I clean and maintain my Countess 3/3FL Automated Cell Counter to help ensure optimal performance?

The following is the recommended procedure for decontaminating the external surface of the Countess™ Automated Cell Counter. Please note that internal decontamination is not possible. •      Wipe the instrument case using a soft, lint-free cloth moistened with 70% alcohol. Wipe the instrument dry immediately after cleaning.
•      Avoid using bleach, as it may leave residue on the case.
•     Ensure that water or other cleaning solutions do not enter the power button, the power inlet, the slide port, or the USB ports.
•     Never pour or spray any liquids directly on the instrument to avoid electrical shock when the instrument is plugged in.


The following is the recommended cleaning process for the Countess Reusable Slide (Cat. No. A25750)

•      Rinse coverslip and Reusable Slide with deionized water to remove debris. A diluted bleach solution or laboratory soap may also be used.
•      Rinse coverslip and Reusable Slide with 70% ethanol to sterilize and reduce drying time.
•      Allow coverslip and Reusable Slide to air dry or use lint-free imaging cloths to speed drying.
•      Reassemble for next sample.

How can I count cells from specific sources, like digested ovarian tumors, using my Countess 3/3FL Automated Cell Counter?

The Countess 3/3FL Automated Cell Counter does not require sample-specific settings in order to count, however users may create customized protocols for individual samples or workflows. Please see Chapter 3 of the user guide.

Countess 3 Automated Cell Counter User Guide (Pub. No. MAN0019566 E.0)

How can I reinstall the software on my Countess 3/3FL Automated Cell Counter if needed?

To reinstall the software on your Countess 3/3FL Automated Cell Counter, take the following steps:

•      Check the exact SW version numbers and make sure that a higher version number is being installed. The Countess instrument will not install a version that has a lower number.
•      Insert a USB drive containing the update file into the Countess with the unit powered on. There is a USB port on the front and back of the instrument and either may be used. The device may take a few seconds to recognize the USB drive. Click on Settings > Software Update > USB > Next > Update. The instrument will update to the software version. Please note this may take many minutes, during which the screen may go black.
•      The USB drive must be FAT32 formatted; verify this before proceeding.
•      The update file must sit on the top level of the USB drive, not within a folder or subfolder.
•      The update file cannot be renamed in any way.
•      The update file cannot be zipped or compressed during distribution.

How can I resolve connectivity issues between my Countess 3/3FL Automated Cell Counter and my computer?

To resolve connectivity issues between your Countess 3/3FL Automated Cell Counter and your computer, take the following steps:

Step 1: Network compatibility
Ensure that the Wi-Fi dongle can support 5 GHz network as they will help get a stronger signal. This can be verified by matching the back of the Wi-Fi dongle with the image below:
Product Image

Step 2: Strength of the network
Ensure that there are enough bars in the Wi-Fi strength icon. Weaker Wi-Fi signal can cause connectivity issues.
Product Image

Check if other instruments with Wi-Fi connectivity are also experiencing similar issues.

Test the Wi-Fi connectivity of the Countess instrument with a mobile hotspot. If mobile hot spot is working, then it would indicate issues with the Wi-Fi signal in the lab.

Step 3: USB port malfunction
The Countess has a front and a rear USB port. Try plugging the USB dongle to the other port and see if that resolves the issue. The USB ports are highlighted as #2 and #7 in the image.
Product Image

Step 4: Thermo Fisher Cloud set up
Please ensure that the instrument is linked to a Thermo Fisher Connect account. The files will not save without a Thermo Fisher Cloud destination. Please set up an account and then try again to connect.

Step 5: Instrument country
Please make sure that the instrument is set to the correct country 'USA & Others' to save the files correctly. To check the country, go to Settings > Instrument Settings > Cloud region

Step 6: File format
Our R&D team found that when naming the files, if there is an extra space after the file name, the instrument would not be able to save the file, and an error screen will appear. Please ensure that there are no extra spaces after the file name.

What should I do if the screen of my Countess 3/3FL Automated Cell Counter is not turning on?

If the screen to your Countess 3/3FL Automated Cell Counter does not turn on, there could be 2 reasons for this mode of failure:

1) Power Supply malfunction: If the power supply is malfunctioning then the instrument will not receive power indicated by the blue light not turning on.

Here are some troubleshooting suggestions:
- Please ensure that there is no damage (cuts, breaks or bends) to the cord and the pins on both ends.
- Test if the power outlet is working with other functional devices.
- Purchase a new Countess™ 3 & 3 FL Power Supply (Cat. No. A48207) and see if the new power supply resolves the issue.

2) Countess hardware issue: Once the power supply being the issue is eliminated then it is likely a Countess hardware failure. In this case, the instrument will have to be sent in for repairs or replaced depending on the service contract. Please contact technical support at Technicalservices@thermofisher.com for further assistance.

How can I update the software of my Countess 3/3FL?

Instructions for updating the Countess 3/3FL software are below:

1. Go to Countess Software Downloads page.
2. Download the latest version of the Countess 3/3 FL software to a computer.
3. Copy the latest countess software file to a USB drive.
4. Insert the USB drive into the Countess 3/3FL instrument (Ensure the .exe update is not hidden within a folder on the USB, as this will prevent a successful update).
5. Press the Setting icon in the lower right corner of the home screen.
Step 5
6. Select "Software Update" from the instrument Settings menu.
Step 6
7. Select the update location (USB or Cloud) and click Next.
8. Select the "Update" button. The update process should only take a minute or two.
Step 8
9. The instrument will automatically restart, and the screen will show when the update is complete.
Step 9

What steps should I follow to troubleshoot common software issues with my Countess 3/3FL Automated Cell Counter?

Tips for troubleshooting a failed Countess 3/3FL software update:

•     Check the exact software version numbers and make sure that a higher version number is being installed. The Countess instrument will not install a version that has a lower number.
•     Insert a USB drive containing the update file into the Countess with the unit powered on. There is a USB port on the front and back of the instrument, and either may be used. The device may take a few seconds to recognize the USB drive. Click on Settings > Software Update > USB > Next > Update. The instrument will update to the software version. Please note this may take many minutes, during which the screen may go black.
•      The USB drive must be FAT32 formatted; verify this before proceeding.
•      The update file must sit within the top level of the USB drive, not within a folder or subfolder.
•      The update file cannot be renamed in any way.
•      The update file cannot be zipped or compressed during distribution.
•      If the update fails, try resetting the instrument. Click Settings > instrument settings > reset instrument.
•      If the update fails, try power-cycling the instrument. Remove the power cable and flip the On/Off switch several times. Allow the instrument to remain powered off for 5 minutes to fully discharge the capacitors. After 5 minutes, plug the instrument back in and reboot.
•      If the above fails to correct the problem, please contact technical support at Technicalservices@thermofisher.com.

How can I reset my Countess 3 Automated Cell Counter to its factory settings?

To reset your Countess 3 Automated Cell Counter to factory settings, use the following steps:

Step 1: Open the settings menu

Step 1 for resetting Countess 3 Automated Cell Counter settings to factory default

Step 2: Go to 'Instrument settings'

Step 2 for resetting Countess 3 Automated Cell Counter settings to factory default

Step 3: Press 'Reset instrument' button

Step 3 for resetting Countess 3 Automated Cell Counter settings to factory default

Why is my WiFi dongle not recognized after being plugged in?

If the dongle is not recognized, please restart the instrument with the dongle plugged in. This can help the instrument properly detect the dongle.

Where can I find information on the warranty and service options for my Countess 3 instrument?

All Countess 3/3 FL Automated Cell Counters come with a standard 1-year warranty. Many customers also choose to purchase a service plan, which extends the standard warranty by two years. Information may be found here under the Services tab.

Countess Automated Cell Counter Services

Where can I find detailed instructions on using software features of my Countess 3 instrument?

Please refer to the following link for many helpful demo videos, found under the Resources tab, covering such topics as "How to save your data" and "How to use on-board calculators."

Countess Automated Cell Counter Resources

Where can I find detailed user manuals for my Countess 3 instrument online?

The user manuals for the Countess 3 and 3FL may be downloaded from the following page, under the Resources tab:

Countess Automated Cell Counter Resources

I noticed your disposable slides come with trypan blue, which is prohibited for importation in my country. Can I order disposable slides without trypan blue?

Yes, you can order the disposable slides without trypan blue here:

Countess Cell Counting Chamber Slide, without Trypan Blue Stain (Cat. No. C10283)

What steps should I follow to install or replace the accessories in my Countess 3 instrument?

Please refer to the following link for helpful demo videos for EVOS light cube installation, reusable slide usage, and more.

Countess Automated Cell Counter Resources

What are the storage and handling recommendations for the consumables used with my Countess 3 Automated Cell Counter?

Storage and handling recommendations may be found in the product manuals for each individual reagent.

For more information about Countess 3 consumables, use the links below.
ReadyCount Stains (Cat. No. A49903, A49904, A49905)

Countess Cell Counting Chamber Slides and Holder, disposable (Cat. Nos. C10228, C10312, C10313, C10314, C10215, T10282, AMEP4745, C10283)

What specific accessories and consumables are compatible with my Countess 3 instrument, and where can I buy them?

The Countess is compatible with easy-to-use ReadyCount reagents for viability counting. We offer both disposable chamber slides and a reusable chamber slide for a greener and more cost-effective option. Additionally, Countess instruments are compatible with the entire line of EVOS light cubes. For more information about these accessories and consumables, use the links below.

ReadyCount Stains (Cat. No. A49903, A49904, A49905)

Countess Cell Counting Chamber Slides and Holder, disposable (Cat. Nos. C10228, C10312, C10313, C10314, C10215, T10282, AMEP4745, C10283)

Countess Reusable Slide and Holder (Cat. Nos. A25750, A48208)

EVOS Light Cubes (Cat. No. AMEP4950, AMEP4951, AMEP4932, AMEP4955, AMEP4956, AMEP4667)

How does my Countess 3 instrument compare with other cell counting products in terms of accuracy, speed, and ease of use?

Automated features like rapid capture, autosave, and built-in calculators add convenience and time savings over hemocytometers or early-generation automated counters. For more information about Countess Automated Cell Counter Features like fast cell counting capabilities, click on the link below.

Countess Automated Cell Counter Features

How can I use my Countess 3 Automated Cell Counter, for cell viability assays, and what are the steps?

Invitrogen ReadyCount Stains are pre-optimized for viability analysis with Countess 3 FL Automated Cell Counter. Simply mix 10 µL of cell sample with 10 µL of stain, pipet the solution into a Countess chamber or reusable slide and insert the slide into the instrument.

Which staining dyes can I use with my Countess 3 FL Automated Cell Counter?

EVOS light cubes, which enable fluorescent detection in the Countess 3 FL Automated Cell Counter, are compatible with a broad range of fluorescent dyes. Refer to our selection guide for an extensive list of compatible dyes. We also offer the ready-to-use ReadyCount Stains for easy viability staining.

How can I count different types of cells, such as yeast cells and PBMCs, with my Countess 3 Automated Cell Counter?

Countess 3 Automated Cell Counters produce accurate and precise counts across a range of cell types, including human and murine peripheral blood mononuclear cells (PBMCs). Cell types and isolated nuclei smaller than 5 microns in diameter may be counted in FL only mode. Fluorescent staining, compatible EVOS light cubes, and update to the latest software version is required.

For more information on EVOS Light Cubes and compatible dyes, please use the following link:

EVOS Cell Imaging System Accessories

What should I do if the software on my Countess 3 Automated Cell Counter freezes during operation?

If your Countess 3 Automated Cell Counter freezes during operation, try power-cycling the instrument. Remove the power cable or flip the On/Off switch several times. This will discharge the capacitors fully. Allow the instrument to remain powered off for 5 minutes. After 5 minutes, plug the instrument back in and reboot. If the above fails to correct the problem, please contact technical support at Technicalservices@thermofisher.com.

What cell counter do you offer that can count live and dead cells, measure cell viability, cell size, has high-resolution touch screen, and comes with reusable slides?
Are there any recommended parameter settings on the Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) for specific cell lines?

No. The Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) parameter settings, especially brightness, can vary between instruments due to slight differences in lamp output and other factors.

What can we do with the FCS file generated by the Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000)?

The Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) FCS files can be uploaded to most flow cytometry software that can accept FCS files, such as FlowJo or the Attune NxT software. Using the flow cytometry software, you may see the distribution of all cells and adjust/fine-tune parameter settings for only the cells of interest or analyze outliers.

Can we use any EVOS Light Cube with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?

With the exception of EVOS Light Cube, White (Cat no. AMEP4672) any of the EVOS Light Cubes for fluorescence detection can be used on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).


For Research Use Only. Not for use in diagnostic procedures.