missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Countess™ 3 FL Automated Cell Counter

Catalog No. AMQAF2000
Encompass
Change view
Click to view available options
Type:
Automated Cell Counter
Automated Cell Counter Starter Package
Warranty:
1-year Standard
3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage)
3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement)
Software:
Countess 3/3 FL Cell Counter Software
Countess 3/3 FL Cell Counter Software with 21 CFR Part 11 Software Module
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Warranty Software
AMQAF2000 Automated Cell Counter 1-year Standard Countess 3/3 FL Cell Counter Software
A49866 Automated Cell Counter Starter Package 1-year Standard Countess 3/3 FL Cell Counter Software
A49892 Automated Cell Counter 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement) Countess 3/3 FL Cell Counter Software
A49893 Automated Cell Counter 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage) Countess 3/3 FL Cell Counter Software
A50299 Automated Cell Counter Starter Package 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement) Countess 3/3 FL Cell Counter Software
A50303 Automated Cell Counter Starter Package 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage) Countess 3/3 FL Cell Counter Software
AMQAF2001 Automated Cell Counter 1-year Standard Countess 3/3 FL Cell Counter Software with 21 CFR Part 11 Software Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. AMQAF2000 Supplier Invitrogen™ Supplier No. AMQAF2000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experience rapid cell counting with the Countess 3 FL Automated Cell Counter. This brightfield and fluorescence cell counter has advanced optics, automated focus and lighting, and AI-based image analysis software for counting cells and nuclei, monitoring fluorescence, and measuring cell viability.

The Countess 3 FL Automated Cell Counter is a benchtop brightfield and fluorescence cell counter with advanced optics, automated focus and lighting, and image analysis software driven by a neural network-based machine-learning algorithm for rapid automated cell counting of primary and immortalized cell lines. The Countess 3 FL counter allows brightfield or fluorescence cell counting with three-channel flexibility (brightfield and two optional fluorescence channels), enabling researchers to count cells and isolated nuclei, monitor fluorescence, examine apoptosis, and measure cell viability.

Features of the Countess 3 FL Automated Cell Counter include:

  • Accurate—highly accurate machine-learning algorithm applied to automated cell counting, eliminates the subjectivity of manual cell counting and user-to-user variability
  • Flexible—accepts up to two EVOS light cubes, user selectable from fifteen fluorescent channels; compatible with both disposable and reusable cell counting chamber slides
  • Fast—counts live and dead cells, measures viability, reports average cell size, calculates percent cell aggregates, and saves data with a single touch, in about 20 seconds
  • Convenient—requires no cleaning, routine maintenance, or calibration; high-resolution capacitive touchscreen and simple user interface provide quick startup and requires minimal training
  • Versatile—perform automated counting of single cells and isolated nuclei for RNA sequencing and gene expression applications in single-cell analysis workflows

The Countess 3 FL Automated Cell Counter works in two simple steps:

  1. Mix 10 μL of sample with 10 μL of trypan blue or other desired reagent, including ReadyCount stain, and pipet into a disposable or reusable Countess chamber slide.
  2. Insert the slide into the instrument. Rapid Capture and Auto-Save modes allow automated cell counting with a single touch. Focus, lighting, and file saving are executed automatically.

Obtain accurate cell counts

The Countess 3 FL Automated Cell Counter uses trypan blue staining combined with auto-focus and lighting driven by a machine-learning image analysis algorithm to obtain accurate cell and viability counts. The measurement range extends from 1 x 104 to 1 x 107 cells/mL (the optimal range is 1 x 105 to 4 x 106 cells/mL), which is broader than that of a hemocytometer.

Simple operation with fluorescent application versatility

Operated using an intuitive user interface, automated cell counting with the Countess 3 FL counter gives you the data you need to prepare your sample for the next passage or experiment and allows you to multiplex with a variety of fluorescent dyes and proteins excited and detected via EVOS light cubes. With both brightfield and fluorescence cell counting options, you can perform cell counts for routine cell culture applications along with more extensive fluorescence analysis of cells such as live vs. dead, nucleated vs. non-nucleated, and transfected vs. untransfected. You no longer need complex, expensive instruments to obtain reliable data.

Transfer data and images to your PC or cloud

Using the USB port on the Countess 3 FL Automated Cell Counter and any USB drive, you can save your cell count results and images and transfer them to your PC. For your convenience, a USB drive and WiFi dongle are supplied with the Countess 3 FL instrument.

Improve downstream single-cell analysis

Ensure accurate single-cell analysis and RNA sequencing results by minimizing cell aggregates and obtaining precise viable cell counts. For optimal single-cell data output, use the percent cell aggregates on the Countess 3 FL to confirm <5% aggregates in the starting sample before single-cell analysis. Staining with fluorescent dyes such as the ReadyCount stains and using the Countess 3 FL-based count mode offers more accurate counts and viability measurements for samples with debris or out-of-focus cells in brightfield. The Countess 3 FL and ReadyCount stains are also recommended for counting isolated nuclei in scATAC-Seq studies. View the single cell and nuclei counting application note for more details.

The Countess 3 FL Automated Cell Counter can be purchased as the instrument (Cat. No. AMQAF2000) or a variety of packages bundled with additional accessories, extended warranties, or software, such as the Countess 3 FL Starter Package (Cat. No. A49866), which includes:

  • Countess FL Automated Cell Counter
  • EVOS GFP and TexasRed light cubes
  • Reusable slide
  • 500 disposable slides with trypan blue

Extended warranty instrument and starter packages

The Countess 3 FL Automated Cell Counter comes with a standard 1-year warranty that covers all costs for shipping, parts, and labor for repairs. Service plans can be bundled with new instrument purchases in an extended warranty package that extends the standard warranty by 2 years for a total of 3. Additional years of coverage can also be purchased to extend the standard 1-year or extended 3-year warranty. The Countess 3 FL instrument and starter packages are offered with two different types of bundled extended warranty packages:

  • 3-year Rapid Exchange (REX) Extended Warranty (1 year standard warranty + 2 years Rapid Exchange, 3 years total exchange entitlement)
    • Replacement shipped out in 1 business day for labs with critical timelines
    • Available with the instrument only (Cat. No. A49892) and the Countess 3 FL Starter Package (Cat. No. A50299)
  • 3-year AB Repair Center (ABRC) Extended Warranty (1 year standard warranty + 2 years extended repair coverage)
    • Instrument repaired and returned in ~3 weeks for regulated labs required to maintain asset tagging consistency (loaner instrument shipped within 1 business day with upgrade to an ABRC Support Plus contract)
    • Available with the instrument only (Cat. No. A49893) and the Countess 3 FL Starter Package (Cat. No. A50303)

Countess 3 FL with 21 CFR Part 11 Software Module (Cat. No. AMQAF2001)

The included Countess 3 21 CFR Part 11 Software Module is for use with the Countess 3 and Countess 3 FL automated cell counters and supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using an SAE administration console. The 21 CFR Part 11 software can also be purchased separately (Cat. No. A51025).

Specifications

Type Automated Cell Counter
Product Line Countess
Camera 5 MP
For Use With (Application) Cell Culture and Analysis Workflows
For Use With (Equipment) EVOS Microscopes, Countess Chamber Slides, and Reusable Slide
Detection Method Brightfield, Fluorescence
Holds 2 samples per slide
Magnification 2.5X
Optics 3 Channels (Brightfield and 2 slots for EVOS™ LED light cubes)
Display LCD (1280 x 800 pixels)
Memory Onboard, USB drive
Software Countess 3/3 FL Cell Counter Software
Dimensions 9 x 9 x 5.5 in. (228.6 x 228.6 x 139.7 mm)
Weight (English) 8 lb.
Weight (Metric) 3.6 kg
Certifications/Compliance RoHS
Content And Storage
  • Countess™ 3 FL Automated Cell Counter, 1 each
  • Power cord with 4 adapters (for US/Canada/Taiwan/Japan, Europe, or UK), 1 each
  • Countess™ Cell Counting Chamber Slides, 50 slides/box
  • Countess™ 3 FL Disposable Slide Holder, 1 each
  • Countess™ 3 FL Reusable Slide Holder, 1 each
  • Countess™ 3 FL Light Cube Removal Tool, 1 each
  • Countess™ 3 USB Drive, 1 each
  • Countess™ 3 FL Automated Cell Counter Quick Reference Card, 1 each
  • Wi-Fi Dongle (Cat. No. A26774), 1 each

Store at room temperature.

Quantity 1 instrument
Shipping Condition Ambient
Warranty 1-year Standard
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
What cell counter do you offer that can count live and dead cells, measure cell viability, cell size, has high-resolution touch screen, and comes with reusable slides?
Why are my cell counts twice as high when using Bright Field (BF) counting on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000), compared to BF+GFP counting, when using the same sample?

The reason for this difference is that the bright field‐only counting assumes that the sample is diluted 1:1 and corrects the result automatically by multiplying it with 2. When you count in the GFP-mode the instrument assumes trypan blue was not used and thus sample not diluted. The total cell concentration displayed after a fluorescent count does not take any dilution into account. Therefore, the results reflect the actual cell concentration in the sample slide, which must be multiplied by any dilution factor present to calculate the original cell concentration.

What is the alternative to TrypanBlue for using it with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000?

The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).

Are there any recommended parameter settings on the Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) for specific cell lines?

No. The Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) parameter settings, especially brightness, can vary between instruments due to slight differences in lamp output and other factors.

What can we do with the FCS file generated by the Countess 3/3 FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000)?

The Countess 3/3FL Automated Cell Counter (Cat. Nos. AMQAF2000, AMQAX2000) FCS files can be uploaded to most flow cytometry software that can accept FCS files, such as FlowJo or the Attune NxT software. Using the flow cytometry software, you may see the distribution of all cells and adjust/fine-tune parameter settings for only the cells of interest or analyze outliers.

Can we use any EVOS Light Cube with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?

With the exception of EVOS Light Cube, White (Cat no. AMEP4672) any of the EVOS Light Cubes for fluorescence detection can be used on the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000).

Can I count nuclei with the Countess 3 Automated Cell Counter (Cat. No. AMQAX2000) or Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?

Yes, with software update version 1.3.1324.552, nuclei can be counted using the Countess 3 FL Automated Cell Counter. We recommend performing FL-based counting. Please refer to this application note for detailed instructions.

What is the optimal cell size and density for accurate cell counting with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000)?

Cells ranging in size from 5 µm to 60 µm and cell suspension density ranging from 1 x 10E5 to 10E6 cells/mL are optimal for counting with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000). 


For Research Use Only. Not for use in diagnostic procedures.