Learn More
Scientific Industries Inc COUNTER DELUXE PILL BALANCE DFS Item
Supplier:  Scientific Industries Inc DRX4C320
Class II pharmacy balance, NTEP certified as legal for trade in prescription weighing. Features a 3,000 drug database capacity and an Rx Verification feature and built with a rugged metal housing and equipped with two RS232 ports. Features receipt printing, three-step pill counting and compounding. Capacity 320g x 0.001g with barcode scanner and backlit LCD.
Catalog No. 501139342
Only null left
Explore available promotions