missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coumarin 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 70°C |
| Color | White |
| Boiling Point | 298°C |
| Quantity | 25 g |
| UN Number | 2811 |
| Formula Weight | 146.15 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Coumarin |
Safety and Handling
| EINECSNumber | (5)-0688 |
| RTECSNumber | GN4200000 |
| TSCA | Yes |