missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 99.0+%, TCI America™

Catalog No. C039525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C039525G 25 g
C0395500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C039525G Supplier TCI America Supplier No. C039525G

Chemical Identifiers

CAS 91-64-5
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006850
InChI Key ZYGHJZDHTFUPRJ-UHFFFAOYSA-N
Synonym coumarin, 2h-chromen-2-one, 2h-1-benzopyran-2-one, cumarin, 1,2-benzopyrone, rattex, tonka bean camphor, coumarinic anhydride, coumarine, benzo-alpha-pyrone
PubChem CID 323
ChEBI CHEBI:28794
IUPAC Name 2H-chromen-2-one
SMILES O=C1OC2=CC=CC=C2C=C1

Specifications

Melting Point 70°C
Color White
Boiling Point 298°C
Quantity 25 g
UN Number 2811
Formula Weight 146.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin
EINECSNumber (5)-0688
RTECSNumber GN4200000
TSCA Yes