missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coumarin 6H 97.0+%, TCI America™
$208.78 - $595.57
Chemical Identifiers
| CAS | 58336-35-9 |
|---|---|
| Molecular Formula | C15H15NO2 |
| Molecular Weight (g/mol) | 241.29 |
| MDL Number | MFCD00227485 |
| InChI Key | MZSOXGPKUOAXNY-UHFFFAOYSA-N |
| Synonym | 2,3,6,7-Tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, 2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin |
| PubChem CID | 94022 |
| SMILES | C1CC2=C3C(=C4C(=C2)C=CC(=O)O4)CCCN3C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C2836200MG
|
TCI America
C2836200MG |
200 mg |
Each for $208.78
|
|
|||||
|
C28361G
|
TCI America
C28361G |
1 g |
Each for $595.57
|
|
|||||
Chemical Identifiers
| 58336-35-9 | |
| 241.29 | |
| MZSOXGPKUOAXNY-UHFFFAOYSA-N | |
| 94022 |
| C15H15NO2 | |
| MFCD00227485 | |
| 2,3,6,7-Tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, 2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin | |
| C1CC2=C3C(=C4C(=C2)C=CC(=O)O4)CCCN3C1 |
Specifications
| 58336-35-9 | |
| Yellow | |
| MFCD00227485 | |
| 2,3,6,7-Tetrahydro-1H-pyrano[2,3-f]pyrido[3,2,1-ij]quinolin-11(5H)-one, 2,3,5,6-1H,4H-Tetrahydroquinolizino[9,9a,1-gh]coumarin | |
| C1CC2=C3C(=C4C(=C2)C=CC(=O)O4)CCCN3C1 | |
| 94022 | |
| ≥97.0% (HPLC) | |
| Coumarin 6H |
| 130°C | |
| C15H15NO2 | |
| 200 mg | |
| MZSOXGPKUOAXNY-UHFFFAOYSA-N | |
| 241.29 | |
| 241.29 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DJ3310000
TSCA : Yes