missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 545T 98.0+%, TCI America™

Catalog No. B4257200MG
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
200 mg
Catalog No. Quantity
B4257200MG 200 mg
1 options

Catalog No. B4257200MG

Supplier: TCI America B4257200MG

Only null left

Chemical Identifiers

CAS 155306-71-1
Molecular Formula C26H26N2O2S
Molecular Weight (g/mol) 430.566
MDL Number MFCD03427187
InChI Key MSDMPJCOOXURQD-UHFFFAOYSA-N
Synonym 10-(2-Benzothiazolyl)-2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one
PubChem CID 5112805
ChEBI CHEBI:51896
SMILES CC1(CCN2CCC(C3=C4C(=CC1=C32)C=C(C(=O)O4)C5=NC6=CC=CC=C6S5)(C)C)C

Specifications

Melting Point 228°C
Quantity 200 mg
Formula Weight 430.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 545T