missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 504T 98.0+%, TCI America™

$177.08 - $605.83

Chemical Identifiers

CAS 113869-06-0
Molecular Formula C22H27NO4
Molecular Weight (g/mol) 369.46
MDL Number MFCD00144013
InChI Key VQQDHBUBOPTRBY-UHFFFAOYSA-N
Synonym coumarin 314t, coumarin 504t, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,5,6,7,11-hexahydro-1h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1,5,7,9 17-tetraene-5-carboxylate, ethyl 2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylate
PubChem CID 1268215
IUPAC Name ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
SMILES CCOC(=O)C1=CC2=CC3=C4N(CCC3(C)C)CCC(C)(C)C4=C2OC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C3045200MG
View Documents
TCI America
C3045200MG
200 mg
Each for $177.08
Only null left
 
C30451G
View Documents
TCI America
C30451G
1 g
Each for $605.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

113869-06-0
369.46
VQQDHBUBOPTRBY-UHFFFAOYSA-N
1268215
CCOC(=O)C1=CC2=CC3=C4N(CCC3(C)C)CCC(C)(C)C4=C2OC1=O
C22H27NO4
MFCD00144013
coumarin 314t, coumarin 504t, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,5,6,7,11-hexahydro-1h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1,5,7,9 17-tetraene-5-carboxylate, ethyl 2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylate
ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate

Specifications

113869-06-0
C22H27NO4
200 mg
VQQDHBUBOPTRBY-UHFFFAOYSA-N
ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
1268215
≥98.0% (HPLC,N)
Coumarin 504T
129°C
MFCD00144013
coumarin 314t, coumarin 504t, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,5,6,7,11-hexahydro-1h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1,5,7,9 17-tetraene-5-carboxylate, ethyl 2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylate
CCOC(=O)C1=CC2=CC3=C4N(CCC3(C)C)CCC(C)(C)C4=C2OC1=O
369.46
369.46
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'