missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 504T 98.0+%, TCI America™

Catalog No. C3045200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3045200MG 200 mg
C30451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3045200MG Supplier TCI America Supplier No. C3045200MG

Chemical Identifiers

CAS 113869-06-0
Molecular Formula C22H27NO4
Molecular Weight (g/mol) 369.46
MDL Number MFCD00144013
InChI Key VQQDHBUBOPTRBY-UHFFFAOYSA-N
Synonym coumarin 314t, coumarin 504t, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 1,1,7,7-tetramethyl-11-oxo-2,3,5,6,7,11-hexahydro-1h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carboxylate, ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo 7.7.1.0 2 ,?.0 1 3 , 1 ? heptadeca-1,5,7,9 17-tetraene-5-carboxylate, ethyl 2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carboxylate
PubChem CID 1268215
IUPAC Name ethyl 10,10,16,16-tetramethyl-4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carboxylate
SMILES CCOC(=O)C1=CC2=CC3=C4N(CCC3(C)C)CCC(C)(C)C4=C2OC1=O

Specifications

Melting Point 129°C
Quantity 200 mg
Formula Weight 369.46
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 504T