missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 337 98.0+%, TCI America™

Catalog No. C3063200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3063200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C3063200MG Supplier TCI America Supplier No. C3063200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55804-68-7
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.30
MDL Number MFCD00051332
InChI Key LGDDFMCJIHJNMK-UHFFFAOYSA-N
Synonym coumarin 337, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carbonitrile, 2,3,6,7-tetrahydro-11-oxo, 2,3,6,7-tetrahydro-11-oxo-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carbonitrile, coumarin 523, pubchem13313, 1h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carbonitrile, 2,3,6,7-tetrahydro-11-oxo, 11-oxo-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinoline-10-carbonitrile, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizine-10-carbonitrile,2,3,6,7-tetrahydro-11-oxo
PubChem CID 72654
IUPAC Name 4-oxo-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraene-5-carbonitrile
SMILES O=C1OC2=C3CCCN4CCCC(C=C2C=C1C#N)=C34

Specifications

Quantity 200 mg
Formula Weight 266.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 337