missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coumarin 314 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 150°C |
| Formula Weight | 313.35 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Quantity | 1 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Coumarin 314 |
Safety and Handling
| TSCA | Yes |