missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 314, 99%, pure, laser grade

CAS: 55804-66-5 | C18H19NO4 | 313.353 g/mol

$94.45 - $94.45

Chemical Identifiers

CAS 55804-66-5
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.353
MDL Number MFCD00051334
InChI Key VMJKUPWQKZFFCX-UHFFFAOYSA-N
Synonym 1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
PubChem CID 72653
ChEBI CHEBI:51940
SMILES CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC405621000
SDS View Documents
Thermo Scientific Chemicals
405621000
100 mg
Each for $94.45
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

55804-66-5
313.353
VMJKUPWQKZFFCX-UHFFFAOYSA-N
72653
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
C18H19NO4
MFCD00051334
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
CHEBI:51940

Specifications

140.0°C to 144.0°C
99%
MFCD00051334
VMJKUPWQKZFFCX-UHFFFAOYSA-N
313.353
CHEBI:51940
99%
Glass bottle
Coumarin 314
55804-66-5
C18H19NO4
1,2,4,5,3H,6H,10H-Tetrahydro-9-carbethoxy[1]benzopyrano(9,9a,1-gh)-quinolizin-10-one
CCOC(=O)C1=CC2=CC3=C4C(=C2OC1=O)CCCN4CCC3
72653
313.34
Pure
100 mg
Videos
SDS
Documents

Documents

RUO – Research Use Only