missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coumarin 314, 99%, pure, laser grade
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 140.0°C to 144.0°C |
| Assay Percent Range | 99% |
| Formula Weight | 313.34 |
| Percent Purity | 99% |
| Grade | Pure |
| Packaging | Glass bottle |
| Quantity | 100 mg |
| Chemical Name or Material | Coumarin 314 |
RUO – Research Use Only