missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 102 97.0+%, TCI America™

Catalog No. C22671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C22671G Supplier TCI America Supplier No. C22671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41267-76-9
Molecular Formula C16H17NO2
Molecular Weight (g/mol) 255.317
MDL Number MFCD00041844
InChI Key XHXMPURWMSJENN-UHFFFAOYSA-N
Synonym coumarin 102, coumarin 480, exciton 480, ccris 4960, 1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 2,3,6,7-tetrahydro-9-methyl, 2,3,5,6-1h,4h-tetrahydro-8-methylquinolazino-9,9a,1-gh coumarin, 2,3,6,7-tetrahydro-9-methyl-1h,5h,11h-1 benzopyrano 6,7,8-ij quinolizin-11-one, 9-methyl-2,3,6,7-tetrahydro-1h,5h,11h-pyrano 2,3-f pyrido 3,2,1-ij quinolin-11-one
PubChem CID 94517
ChEBI CHEBI:51774
SMILES CC1=CC(=O)OC2=C3CCCN4C3=C(CCC4)C=C12

Specifications

Melting Point 151°C
Quantity 1 g
Formula Weight 255.32
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 102
RTECSNumber DJ3320000
TSCA Yes