missing translation for 'onlineSavingsMsg'
Learn More

Coumalic Acid 97.0+%, TCI America™

Catalog No. C03915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03915G 5 g
C039125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03915G Supplier TCI America Supplier No. C03915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 500-05-0
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00006644
InChI Key ORGPJDKNYMVLFL-UHFFFAOYSA-N
Synonym coumalic acid, 2-oxo-2h-pyran-5-carboxylic acid, cumalic acid, 2h-pyran-5-carboxylic acid, 2-oxo, 2-oxopyran-5-carboxylic acid, unii-ob1jpy343g, 2-pyrone-5-carboxylic acid, alpha-pyrone-5-carboxylic acid, .alpha.-pyrone-5-carboxylic acid, ob1jpy343g
PubChem CID 68141
IUPAC Name 2-oxo-2H-pyran-5-carboxylic acid
SMILES OC(=O)C1=COC(=O)C=C1

Specifications

Melting Point 204°C
Boiling Point 218°C
Quantity 5 g
Formula Weight 140.09
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Coumalic Acid