Learn More
Coulter™ IsoFlow™ Sheath Fluid
Description
Includes
- Sodium chloride, which allows diluent to become an electrolyte capable of conducting electrical current in an electronic analyzer and, along with phosphate salts, provides buffer for pH balance
- Buffering agent containing an anticoagulant, EDTA, bacteriostatic agent, and sodium fluoride
Manufactured for low particulate and fluorescence background to ensure superior signal to noise ratio measurements during the analysis process
Specifications
Specifications
| Quantity | 4 x 1.8 L |
| Product Type | Sheath Fluid |
| Includes | 4 x 1.8 L IsoFlow™ sheath fluid, sodium chloride and phosphate salts |
| Certifications/Compliance | IVD, CE, FCM |
| Format | Liquid |